accessibility menu, dialog, popup

UserName

CH3OClBr

CAS RN 34911-51-8

IUPAC Name: 2-bromo-1-(3-chlorophenyl)propan-1-one
Synonyms: 2-bromo-1-(3-chlorophenyl)propan-1-one
Molecular Weight (g/mol): 247.52
Molecular Formula: C9H8BrClO
InChi Key: OFNMQTRHMBQQEA-UHFFFAOYNA-N
SMILES: CC(Br)C(=O)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results