accessibility menu, dialog, popup

UserName

CAS RN 349-46-2

NH2(R)SS(R)NH2HOOOHO

CAS RN 349-46-2

IUPAC Name: (2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Synonyms: L-cystine β,β'-dithioalanine, L- b,b'-dithiodialanine gelucystine L-dicysteine 1-cystine L cystine oxidized L-cysteine copper cystinate 3,3'-dithiobis
Molecular Weight (g/mol): 240.29
Molecular Formula: C6H12N2O4S2
InChi Key: LEVWYRKDKASIDU-IMJSIDKUSA-N
SMILES: N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results