accessibility menu, dialog, popup

UserName

CAS RN 348134-09-8

IUPAC Name: 6-chloro-2-(1,4-diazepan-1-yl)-1,3-benzothiazole
Synonyms: 6-chloro-2-(1,4-diazepan-1-yl)-1,3-benzothiazole
Molecular Weight (g/mol): 267.78
Molecular Formula: C12H14ClN3S
InChi Key: UZLJYDCBJULBQU-UHFFFAOYSA-N
SMILES: ClC1=CC=C2N=C(SC2=C1)N1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results