accessibility menu, dialog, popup

UserName

CH3CH3H3CSOOH2N

CAS RN 34813-49-5

IUPAC Name: 2-methylpropane-2-sulfonamide
Synonyms: 2-methylpropane-2-sulfonamide
Molecular Weight (g/mol): 137.2
Molecular Formula: C4H11NO2S
InChi Key: GWJSQKNYHPYZRN-UHFFFAOYSA-N
SMILES: CC(C)(C)S(N)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results