CAS RN 348-26-5
IUPAC Name:
5-fluoro-1H-indazole
Synonyms:
5-fluoro-1H-indazole
Molecular Weight (g/mol):
136.13
Molecular Formula:
C7H5FN2
InChi Key:
LIWIWTHSKJBYDW-UHFFFAOYSA-N
SMILES:
FC1=CC=C2NN=CC2=C1
1
–
1
of
1
results
1
Promotions Available
5-Fluoro-1H-indazole, 98%
5-Fluoro-1H-indazole, 98%
CAS: 348-26-5 Molecular Formula: C7H5FN2 Molecular Weight (g/mol): 136.129 MDL Number: MFCD04972877 InChI Key: LIWIWTHSKJBYDW-UHFFFAOYSA-N Synonym: 5-fluoroindazole,5-fluoro-2h-indazole,1h-indazole, 5-fluoro,chembl16076,5-fluoranyl-1h-indazole,2h-indazole, 5-fluoro,acmc-209zy1,indazole, 5-fluoro,5-fluoro-1h-indazole PubChem CID: 17842486 IUPAC Name: 5-fluoro-1H-indazole SMILES: C1=CC2=C(C=C1F)C=NN2