accessibility menu, dialog, popup

UserName

CAS RN 34722-01-5

H3CS

CAS RN 34722-01-5

IUPAC Name: 3-butylthiophene
Synonyms: 3-butylthiophene
Molecular Weight (g/mol): 140.24
Molecular Formula: C8H12S
InChi Key: KPOCSQCZXMATFR-UHFFFAOYSA-N
SMILES: CCCCC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results