accessibility menu, dialog, popup

UserName

CAS RN 347-84-2

IUPAC Name: 1-(4-fluorophenyl)-2-phenylethan-1-one
Synonyms: 1-(4-fluorophenyl)-2-phenylethanone
Molecular Weight (g/mol): 214.24
Molecular Formula: C14H11FO
InChi Key: YFYKGCQUWKAFLW-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results