accessibility menu, dialog, popup

UserName

CH3SiClClH3C

CAS RN 3449-28-3

IUPAC Name: dibutyldichlorosilane
Synonyms: dibutyldichlorosilane
Molecular Weight (g/mol): 213.22
Molecular Formula: C8H18Cl2Si
InChi Key: NJKDOKBDBHYMAH-UHFFFAOYSA-N
SMILES: CCCC[Si](Cl)(Cl)CCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results