accessibility menu, dialog, popup

UserName

H3CNH3COOON

CAS RN 34372-72-0

IUPAC Name: 7'-(dibenzylamino)-3'-(diethylamino)-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 7'-(dibenzylamino)-3'-(diethylamino)spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 566.7
Molecular Formula: C38H34N2O3
InChi Key: DXYZIGZCEVJFIX-UHFFFAOYNA-N
SMILES: CCN(CC)C1=CC=C2C(OC3=CC=C(C=C3C22OC(=O)C3=CC=CC=C23)N(CC2=CC=CC=C2)CC2=CC=CC=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results