CAS RN 3435-28-7
IUPAC Name:
6-methylpyrimidin-4-amine
Synonyms:
6-methyl-4-pyrimidinamine
Molecular Weight (g/mol):
109.13
Molecular Formula:
C5H7N3
InChi Key:
LAERIBHKDNBVOO-UHFFFAOYSA-N
SMILES:
CC1=CC(N)=NC=N1
1
–
1
of
1
results
1
Promotions Available
4-Amino-6-methylpyrimidine, 95%
4-Amino-6-methylpyrimidine, 95%
CAS: 3435-28-7 Molecular Formula: C5H7N3 Molecular Weight (g/mol): 109.132 MDL Number: MFCD01863677 InChI Key: LAERIBHKDNBVOO-UHFFFAOYSA-N Synonym: 4-amino-6-methylpyrimidine,6-methyl-4-pyrimidinamine,4-pyrimidinamine, 6-methyl,6-methylpyrimidin-4-ylamine,4-pyrimidinamine, 6-methyl-9ci,4-methyl-6-aminopyrimidine,4-pyrimidinamine,6-methyl,6-methylpyrimidine-4-ylamine,6-methyl-4-amine-pyrimidine,6-methyl-4-pyrimidinamine # PubChem CID: 572852 IUPAC Name: 6-methylpyrimidin-4-amine SMILES: CC1=CC(=NC=N1)N