accessibility menu, dialog, popup

UserName

HO

CAS RN 34281-92-0

IUPAC Name: 2-phenylcyclohexan-1-ol
Synonyms: cyclohexanol, 2-phenyl-
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: AAIBYZBZXNWTPP-UHFFFAOYNA-N
SMILES: OC1CCCCC1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results