accessibility menu, dialog, popup

UserName

CAS RN 34270-90-1

IOI

CAS RN 34270-90-1

IUPAC Name: 1-iodo-2-(2-iodoethoxy)ethane
Synonyms: 1-iodo-2-(2-iodoethoxy)ethane
Molecular Weight (g/mol): 325.92
Molecular Formula: C4H8I2O
InChi Key: XWYDQETVMJZUOJ-UHFFFAOYSA-N
SMILES: ICCOCCI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results