accessibility menu, dialog, popup

UserName

CH3CH3H3C(E)

CAS RN 3404-75-9

IUPAC Name: (2E)-3-methylhept-2-ene
Synonyms: (2E)-3-methylhept-2-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: OFKLSPUVNMOIJB-VMPITWQZSA-N
SMILES: CCCC\C(C)=C\C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results