accessibility menu, dialog, popup

UserName

CAS RN 33984-50-8

IUPAC Name: N-[(4-methoxyphenyl)methyl]-7-nitro-2,1,3-benzoxadiazol-4-amine
Synonyms: N-[(4-methoxyphenyl)methyl]-7-nitro-2,1,3-benzoxadiazol-4-amine
Molecular Weight (g/mol): 300.27
Molecular Formula: C14H12N4O4
InChi Key: IENONFJSMWUIQQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNC2=CC=C(C3=NON=C23)[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results