accessibility menu, dialog, popup

UserName

H3COOOOOOCH3

CAS RN 33926-25-9

IUPAC Name: 4-ethoxyphenyl 4-[(ethoxycarbonyl)oxy]benzoate
Synonyms: 4-ethoxyphenyl 4-[(ethoxycarbonyl)oxy]benzoate
Molecular Weight (g/mol): 330.34
Molecular Formula: C18H18O6
InChi Key: NRRSSIQKLMBWJB-UHFFFAOYSA-N
SMILES: CCOC(=O)OC1=CC=C(C=C1)C(=O)OC1=CC=C(OCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results