accessibility menu, dialog, popup

UserName

H3CNHNOHCH3(E/Z)

CAS RN 339068-25-6

IUPAC Name: N'-(4-butyl-2-methylphenyl)-N-hydroxymethanimidamide
Synonyms: N'-(4-butyl-2-methylphenyl)-N-hydroxymethanimidamide
Molecular Weight (g/mol): 206.29
Molecular Formula: C12H18N2O
InChi Key: LYNOGBKNFIHKLE-UHFFFAOYSA-N
SMILES: CCCCC1=CC(C)=C(C=C1)N=CNO
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results