accessibility menu, dialog, popup

UserName

CAS RN 3382-18-1

IUPAC Name: 6,7-dimethoxy-3,4-dihydroisoquinoline
Synonyms: 6,7-dimethoxy-3,4-dihydroisoquinoline
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: NSLJVQUDZCZJLK-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=NCC2)C=C1OC
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)