accessibility menu, dialog, popup

UserName

CAS RN 3363-97-1

OOOO

CAS RN 3363-97-1

IUPAC Name: 1-[4-(2-oxo-2-phenylacetyl)phenyl]-2-phenylethane-1,2-dione
Synonyms: 1-[4-(2-oxo-2-phenylacetyl)phenyl]-2-phenylethane-1,2-dione
Molecular Weight (g/mol): 342.35
Molecular Formula: C22H14O4
InChi Key: FUEGWHHUYNHBNI-UHFFFAOYSA-N
SMILES: O=C(C(=O)C1=CC=C(C=C1)C(=O)C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results