accessibility menu, dialog, popup

UserName

NaNaOSOOOSOOO

CAS RN 33513-44-9

IUPAC Name: disodium 4-formylbenzene-1,3-disulfonate
Synonyms: disodium 4-formylbenzene-1,3-disulfonate
Molecular Weight (g/mol): 310.2
Molecular Formula: C7H4Na2O7S2
InChi Key: UUKHCUPMVISNFW-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=O)C(=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results