accessibility menu, dialog, popup

UserName

CAS RN 335-95-5

NaOOFFFFFFFFFFFFFFF

CAS RN 335-95-5

IUPAC Name: sodium pentadecafluorooctanoate
Synonyms: sodium perfluorooctanoate
Molecular Weight (g/mol): 436.05
Molecular Formula: C8F15NaO2
InChi Key: LWHQXUODFPPQTL-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results