accessibility menu, dialog, popup

UserName

H3CCH3(S)OOCH3NHH3CHCl

CAS RN 3339-44-4

IUPAC Name: methyl (2S)-3-methyl-2-(methylamino)butanoate hydrochloride
Synonyms: methyl (2S)-3-methyl-2-(methylamino)butanoate hydrochloride|N-ME-VAL-OME HCL
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: DLLHGHUMLSLOID-RGMNGODLSA-N
SMILES: Cl.CN[C@@H](C(C)C)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)