accessibility menu, dialog, popup

UserName

CAS RN 33332-25-1

IUPAC Name: methyl 5-chloropyrazine-2-carboxylate
Synonyms: methyl 5-chloropyrazine-2-carboxylate
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: CVVMLRFXZPKILB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(Cl)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results