CAS RN 3333-15-1
IUPAC Name:
2,3-diphenylpropanoic acid
Synonyms:
2,3-diphenylpropanoic acid
Molecular Weight (g/mol):
226.28
Molecular Formula:
C15H14O2
InChi Key:
PCRICPYPVZKEBZ-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2,3-Diphenylpropionic Acid 98.0+%, TCI America™
CAS: 3333-15-1 Molecular Formula: C15H14O2 Molecular Weight (g/mol): 226.28 MDL Number: MFCD00044405 InChI Key: PCRICPYPVZKEBZ-UHFFFAOYNA-N PubChem CID: 18744 IUPAC Name: 2,3-diphenylpropanoic acid SMILES: OC(=O)C(CC1=CC=CC=C1)C1=CC=CC=C1