accessibility menu, dialog, popup

UserName

CAS RN 33225-74-0

NOHONOO

CAS RN 33225-74-0

IUPAC Name: 2-nitropyridine-4-carboxylic acid
Synonyms: 2-nitropyridine-4-carboxylic acid
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: SXERXZLYZFAKBX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results