accessibility menu, dialog, popup

UserName

AgON

CAS RN 3315-16-0

IUPAC Name: silver(1+) cyanate
Synonyms: silver cyanate
Molecular Weight (g/mol): 149.89
Molecular Formula: CAgNO
InChi Key: DWBPIWPCOSHWCK-UHFFFAOYSA-M
SMILES: [Ag+].[O-]C#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results