accessibility menu, dialog, popup

UserName

CAS RN 33098-21-4

IUPAC Name: 4,5-dichloro-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one
Synonyms: 4,5-dichloro-2-(4-methylphenyl)pyridazin-3-one
Molecular Weight (g/mol): 255.1
Molecular Formula: C11H8Cl2N2O
InChi Key: OWPCMQKRJFNLHF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N1N=CC(Cl)=C(Cl)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)