accessibility menu, dialog, popup

UserName

H3CCH3(S)NHO(S)CH3NH2OHO

CAS RN 3303-34-2

IUPAC Name: (2S)-2-[(2S)-2-aminopropanamido]-4-methylpentanoic acid
Synonyms: Ala-Leu
Molecular Weight (g/mol): 202.25
Molecular Formula: C9H18N2O3
InChi Key: RDIKFPRVLJLMER-WZTWBHKBNA-N
SMILES: CC(C)C[C@H](NC(=O)[C@H](C)N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)