accessibility menu, dialog, popup

UserName

OOFFFFFF

CAS RN 32890-87-2

IUPAC Name: 2,4-bis(trifluoromethyl)benzoate
Synonyms: 2,4-bis(trifluoromethyl)benzoate
Molecular Weight (g/mol): 257.11
Molecular Formula: C9H3F6O2
InChi Key: JTOIZLCQNWWDCN-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)