accessibility menu, dialog, popup

UserName

CAS RN 32833-96-8

CH3NH3COOHO

CAS RN 32833-96-8

IUPAC Name: (dimethylcarbamoyl)formic acid
Synonyms: (dimethylcarbamoyl)formic acid
Molecular Weight (g/mol): 117.1
Molecular Formula: C4H7NO3
InChi Key: YKFGLGXRUVEMNF-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results