CAS RN 3230-94-2
CAS RN 3230-94-2
IUPAC Name:
(4S)-4-amino-4-carboxybutan-1-aminium (2S)-2-amino-3-carboxypropanoate
Synonyms:
(4S)-4-amino-4-carboxybutan-1-aminium (+)-aspartate
Molecular Weight (g/mol):
265.27
Molecular Formula:
C9H19N3O6
InChi Key:
IXUZXIMQZIMPSQ-ZBRNBAAYSA-N
SMILES:
N[C@@H](CCC[NH3+])C(O)=O.N[C@@H](CC(O)=O)C([O-])=O
1
–
1
of
1
results
1
L-Ornithine L-Aspartate 96.0+%, TCI America™
CAS: 3230-94-2 Molecular Formula: C9H19N3O6 Molecular Weight (g/mol): 265.27 MDL Number: MFCD00058084 InChI Key: IXUZXIMQZIMPSQ-ZBRNBAAYSA-N PubChem CID: 10220941 IUPAC Name: (4S)-4-amino-4-carboxybutan-1-aminium (2S)-2-amino-3-carboxypropanoate SMILES: N[C@@H](CCC[NH3+])C(O)=O.N[C@@H](CC(O)=O)C([O-])=O