accessibility menu, dialog, popup

UserName

BrOHN

CAS RN 320734-35-8

IUPAC Name: 7-bromo-2,3-dihydro-1H-indol-2-one
Synonyms: 7-bromo-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: WSUWXWBRIBGIQT-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1NC(=O)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results