accessibility menu, dialog, popup

UserName

CAS RN 31972-52-8

H3CCH3H3COOHNONHOHO

CAS RN 31972-52-8

IUPAC Name: 2-(2-{[(tert-butoxy)carbonyl]amino}acetamido)acetic acid
Synonyms: {2-[(tert-butoxycarbonyl)amino]acetamido}acetic acid
Molecular Weight (g/mol): 232.24
Molecular Formula: C9H16N2O5
InChi Key: HWBAHOVOSOAFLE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results