accessibility menu, dialog, popup

UserName

CAS RN 318469-22-6

CH3NNFFFHOO

CAS RN 318469-22-6

IUPAC Name: [1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Synonyms: [1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazol-4-yl]methanol
Molecular Weight (g/mol): 272.23
Molecular Formula: C12H11F3N2O2
InChi Key: TVYDMZWMEOZWHY-UHFFFAOYSA-N
SMILES: CN1N=C(C(CO)=C1OC1=CC=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)