accessibility menu, dialog, popup

UserName

CAS RN 3160-91-6

IUPAC Name: N''-(morpholine-4-carboximidoyl)guanidine hydrochloridyl
Synonyms: moroxydine hydrochloridyl
Molecular Weight (g/mol): 206.65
Molecular Formula: C6H13ClN5O
InChi Key: LEUPELREAARNLR-UHFFFAOYSA-N
SMILES: [Cl].NC(N)=NC(=N)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results