accessibility menu, dialog, popup

UserName

BrSBr

CAS RN 31574-87-5

IUPAC Name: 4,12-dibromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 4,12-dibromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 342.05
Molecular Formula: C12H6Br2S
InChi Key: WNEXSUAHKVAPFK-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(SC3=C2C=C(Br)C=C3)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results