CAS RN 31462-59-6
CAS RN 31462-59-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pyrimidine-4-carboxylic Acid 98.0+%, TCI America™
CAS: 31462-59-6 Molecular Formula: C5H4N2O2 Molecular Weight (g/mol): 124.099 MDL Number: MFCD00129737 InChI Key: YPOXGDJGKBXRFP-UHFFFAOYSA-N Synonym: 4-pyrimidinecarboxylic acid,pubchem5297,acmc-1cpdb,4-pyrimidine-carboxylicacid,4-carboxy pyrimidine,pyrimidine 4-carboxylic acid,ksc223e8f,pyrimidine-4-carboxylic acid,pyrimidin-4-yl-carboxylic acid,4-pyrimidinecarboxylic acid 6ci,8ci,9ci PubChem CID: 169306 IUPAC Name: pyrimidine-4-carboxylic acid SMILES: C1=CN=CN=C1C(=O)O