accessibility menu, dialog, popup

UserName

BrSBrBr

CAS RN 3141-24-0

IUPAC Name: 2,3,5-tribromothiophene
Synonyms: thiophene, 2,3,5-tribromo-
Molecular Weight (g/mol): 320.82
Molecular Formula: C4HBr3S
InChi Key: SKDNDSLDRLEELJ-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C(Br)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results