accessibility menu, dialog, popup

UserName

CAS RN 312693-72-4

IUPAC Name: 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one hydrate
Synonyms: 2-deoxyguanosine hydrate
Molecular Weight (g/mol): 285.26
Molecular Formula: C10H15N5O5
InChi Key: LZSCQUCOIRGCEJ-VDPVQVSMNA-N
SMILES: O.NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)