accessibility menu, dialog, popup

UserName

H3COO

CAS RN 3121-71-9

IUPAC Name: naphthalen-1-yl propanoate
Synonyms: naphthalen-1-yl propanoate
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: FRXPDEZCWCPLIH-UHFFFAOYSA-N
SMILES: CCC(=O)OC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results