CAS RN 3085-30-1
CAS RN 3085-30-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Aluminum n-butoxide, 90+%
CAS: 3085-30-1 Molecular Formula: C12H30AlO3 Molecular Weight (g/mol): 249.351 MDL Number: MFCD00015237 InChI Key: YGVBJFIUURLJDC-UHFFFAOYSA-N Synonym: aluminum iii n-butoxide,aluminum; butan-1-ol PubChem CID: 50933522 IUPAC Name: aluminum;butan-1-ol SMILES: CCCCO.CCCCO.CCCCO.[Al]