accessibility menu, dialog, popup

UserName

CAS RN 3073-77-6

NH2NNNOO

CAS RN 3073-77-6

IUPAC Name: 5-nitropyrimidin-2-amine
Synonyms: 5-nitropyrimidin-2-amine
Molecular Weight (g/mol): 140.1
Molecular Formula: C4H4N4O2
InChi Key: SSHFCFRJYJIJDV-UHFFFAOYSA-N
SMILES: NC1=NC=C(C=N1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results