accessibility menu, dialog, popup

UserName

H2CCH2

CAS RN 3070-53-9

IUPAC Name: hepta-1,6-diene
Synonyms: 1,6-heptadiene
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: GEAWFZNTIFJMHR-UHFFFAOYSA-N
SMILES: C=CCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results