accessibility menu, dialog, popup

UserName

H3COOCH3CH3

CAS RN 30673-38-2

IUPAC Name: 2-methylpropyl decanoate
Synonyms: 2-methylpropyl decanoate
Molecular Weight (g/mol): 228.38
Molecular Formula: C14H28O2
InChi Key: AXTPGRJJIGEOOY-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results