accessibility menu, dialog, popup

UserName

CAS RN 306-37-6

HHClClCH3HNNHH3C

CAS RN 306-37-6

IUPAC Name: dihydrogen 1,2-dimethylhydrazine dichloride
Synonyms: dihydrogen 1,2-dimethylhydrazine dichloride
Molecular Weight (g/mol): 133.02
Molecular Formula: C2H10Cl2N2
InChi Key: WLDRFJDGKMTPPV-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CNNC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results