CAS RN 30536-19-7
IUPAC Name:
5-chloro-2,1,3-benzothiadiazol-4-amine
Synonyms:
5-chloro-2,1,3-benzothiadiazol-4-amine
Molecular Weight (g/mol):
185.63
Molecular Formula:
C6H4ClN3S
InChi Key:
MURNIACGGUSMAP-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C=CC2=NSN=C12
1
–
1
of
1
results
1
4-Amino-5-chloro-2,1,3-benzothiadiazole 97.0+%, TCI America™
CAS: 30536-19-7 Molecular Formula: C6H4ClN3S Molecular Weight (g/mol): 185.629 MDL Number: MFCD00465908 InChI Key: MURNIACGGUSMAP-UHFFFAOYSA-N Synonym: 4-Amino-5-chloropiazthiole PubChem CID: 771620 IUPAC Name: 5-chloro-2,1,3-benzothiadiazol-4-amine SMILES: C1=CC2=NSN=C2C(=C1Cl)N