CAS RN 304445-49-6
IUPAC Name:
1-(4-bromo-3-fluorophenyl)ethan-1-one
Synonyms:
1-(4-bromo-3-fluorophenyl)ethanone
Molecular Weight (g/mol):
217.04
Molecular Formula:
C8H6BrFO
InChi Key:
VCTWSAITPPCBHI-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(F)=C(Br)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Bromo-3-fluoroacetophenone, 96%, Thermo Scientific Chemicals
4-Bromo-3-fluoroacetophenone, 96%, Thermo Scientific Chemicals
CAS: 304445-49-6 Molecular Formula: C8H6BrFO Molecular Weight (g/mol): 217.037 MDL Number: MFCD00466241 InChI Key: VCTWSAITPPCBHI-UHFFFAOYSA-N Synonym: 1-4-bromo-3-fluorophenyl ethanone,4'-bromo-3'-fluoroacetophenone,3-fluoro-4-bromo-acetophenone,1-4-bromo-3-fluorophenyl ethan-1-one,4-bromo-3-fluoroacetophenone,4-bromo-3-fluoro-acetophenone,ksc915o9p,3'-fluoro-4'-bromo-acetophenone,1-4-bromo-3-fluoro-phenyl-ethanone PubChem CID: 22831914 IUPAC Name: 1-(4-bromo-3-fluorophenyl)ethanone SMILES: CC(=O)C1=CC(=C(C=C1)Br)F