accessibility menu, dialog, popup

UserName

CAS RN 303734-52-3

IUPAC Name: 2-bromo-3-(2-ethylhexyl)thiophene
Synonyms: 2-bromo-3-(2-ethylhexyl)thiophene
Molecular Weight (g/mol): 275.25
Molecular Formula: C12H19BrS
InChi Key: VWIAAXXJIUXUEC-UHFFFAOYNA-N
SMILES: CCCCC(CC)CC1=C(Br)SC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results