accessibility menu, dialog, popup

UserName

CAS RN 3032-92-6

IUPAC Name: 4-ethynylbenzonitrile
Synonyms: 4-ethynylbenzonitrile
Molecular Weight (g/mol): 127.15
Molecular Formula: C9H5N
InChi Key: LAGNMUUUMQJXBF-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)