accessibility menu, dialog, popup

UserName

SNOCl

CAS RN 30216-57-0

IUPAC Name: 1,3-thiazole-2-carbonyl chloride
Synonyms: 1,3-thiazole-2-carbonyl chloride
Molecular Weight (g/mol): 147.58
Molecular Formula: C4H2ClNOS
InChi Key: HKZCFGQSWLWNQA-UHFFFAOYSA-N
SMILES: ClC(=O)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results