accessibility menu, dialog, popup

UserName

H3COO(R)(S)(R)ClO(R)H3C(S)(S)H3CH3CO

CAS RN 302-22-7

IUPAC Name: (1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-5-chloro-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Synonyms: Cero chlormadinonu menstridyl lutinyl verton bovisynchron clordion chlormadinone acetate traslan gestafortin
Molecular Weight (g/mol): 404.93
Molecular Formula: C23H29ClO4
InChi Key: QMBJSIBWORFWQT-DFXBJWIESA-N
SMILES: CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results